

cis-1,3-Cyclohexanediamine ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:26772-34-9
:
Formula:C₆H₁₄N₂
:114.19 g/mol
Boiling Pt:198°C
UN:2735
ADR:8,III
:GHS05
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together