
2-(4-Chlorophenyl)-3-methylbutyric acid ≥98.0% (by titrimetric analysis)
Catalog # TCIAC2741-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 97.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:87.0 to 91.0 deg-C
Specification Test Results
About this item
:
:212.67 g/mol
EINECS:217-934-1
:GHS07
CAS Number:2012-74-0
Formula:C₁₁H₁₃ClO₂
MDL Number:MFCD00037763