
Chloro[(tri-tert-butylphosphine)-2-(2-aminobiphenyl)]palladium(II) ≥98.0% (by titrimetric analysis)
Catalog # TCIAC2734-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Purity (Chelometric Titration):min. 98.0 %
Specification Test Results
About this item
:
:512.41 g/mol
:GHS07
CAS Number:1375325-71-5
Formula:C₂₄H₃₇ClNPPd
MDL Number:MFCD21608496