Order Entry
Italy
ContactUsLinkComponent
4H-Cyclopenta[1,2-b:5,4-b']dithiophen-4-one ≥98.0% (by GC)
4H-Cyclopenta[1,2-b:5,4-b']dithiophen-4-one ≥98.0% (by GC)
Catalog # TCIAC2715-200MG
Supplier:  TCI
CAS Number:  
4H-Cyclopenta[1,2-b:5,4-b']dithiophen-4-one ≥98.0% (by GC)
Catalog # TCIAC2715-200MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    200 mg
  • Seq:
    2530

Specifications

Specification Test Results

  • Appearance:
    Orange to Amber to Dark red powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    134.0 to 138.0 deg-C

Specification Test Results

About this item

 :
 :192.25 g/mol
CAS Number:25796-77-4
Formula:C₉H₄OS₂
MDL Number:MFCD16619150