2-(((Benzyloxy)carbonyl)amino)-2-hydroxyacetic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:56538-57-9
:
Formula:C₁₀H₁₁NO₅
MDL Number:MFCD02179279
:225.2 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
Frequently Bought Together