Methoxy(cyclooctadiene)iridium(I) dimer
Supplier: TCI
Certificates
About this item
CAS Number:12148-71-9
:
Formula:C₁₈H₃₀Ir₂O₂
MDL Number:MFCD08459360
:662.87 g/mol
Storage Temperature:Freezer
Specification Test Results
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Elemental analysis (Carbon):31.50 to 34.00 %
Specification Test Results
Related Information
Frequently Bought Together