Clonixin ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
Synonyms: 2-[(3-Chloro-2-methylphenyl)amino]nicotinic Acid, 2-(3-Chloro-2-methylanilino)pyridine-3-carboxylic acid, 2-(3-Chloro-o-toluidino)nicotinic Acid
CAS Number:17737-65-4
:
Formula:C₁₃H₁₁ClN₂O₂
MDL Number:MFCD00864292
:262.69 g/mol
Merck Index:14,02392
EINECS:241-730-1
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:240.0 to 244.0 deg-C
Specification Test Results
Frequently Bought Together