Order Entry
Italy
ContactUsLinkComponent
2-(Chloromethyl)-1,2-epoxypropane ≥98.0% (by GC)
Flammable
Corrosive
2-(Chloromethyl)-1,2-epoxypropane ≥98.0% (by GC)
Catalog # TCIAC2584-5G
Supplier:  TCI
CAS Number:  
Flammable
Corrosive
2-(Chloromethyl)-1,2-epoxypropane ≥98.0% (by GC)
Catalog # TCIAC2584-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :106.55 g/mol
Boiling Pt:122°C
Flash Pt:30°C
EINECS:209-916-7
UN:2920
ADR:8,II
 :GHS02
 :GHS05
CAS Number:598-09-4
Formula:C₄H₇ClO
MDL Number:MFCD00052487