2-Chloro-7H-purin-6-amine ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:1839-18-5
:
Formula:C₅H₄ClN₅
MDL Number:MFCD00037925
:169.57 g/mol
Specification Test Results
- Appearance:White to Light yellow to Dark green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together