Order Entry
Italy
ContactUsLinkComponent
Crystal violet for biochemical research
Corrosive
Irritant
Health
Environment
Crystal violet for biochemical research
Catalog # TCIAC2460-5G
Supplier:  TCI
CAS Number:  
undefined
Corrosive
Irritant
Health
Environment
Crystal violet for biochemical research
Catalog # TCIAC2460-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Green to Dark green powder to crystal
  • Absorbance:
    min. 0.300 (2mg/L, 588.0 to 592.0 nm) (calcd.on dried substance)
  • Cell staining test:
    to pass test

Specification Test Results

About this item

 :
 :407.99 g/mol
Merck Index:14,04395
Melting Pt:211°C
EINECS:208-953-6
UN:3077
ADR:9,III
 :GHS05
 :GHS07
 :GHS08
 :GHS09
CAS Number:548-62-9
Formula:C₂₅H₃₀ClN₃
MDL Number:MFCD00011750