Order Entry
Italy
ContactUsLinkComponent
[12]Cycloparaphenylene ≥90.0% (by HPLC)
[12]Cycloparaphenylene ≥90.0% (by HPLC)
Catalog # TCIAC2449-10MG
Supplier:  TCI
CAS Number:  
[12]Cycloparaphenylene ≥90.0% (by HPLC)
Catalog # TCIAC2449-10MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Seq:
    2275

Specifications

Specification Test Results

  • Appearance:
    Light orange to Yellow to Green powder to crystal
  • Purity (HPLC):
    min. 90.0 area%

Specification Test Results

About this item

 :
 :913.18 g/mol
CAS Number:1092522-75-2
Formula:C₇₂H₄₈