Order Entry
Italy
ContactUsLinkComponent
2-Chlorophenyl trifluoromethanesulphonate ≥97.0% (by GC)
Irritant
2-Chlorophenyl trifluoromethanesulphonate ≥97.0% (by GC)
Catalog # TCIAC2326-5G
Supplier:  TCI
CAS Number:  
Irritant
2-Chlorophenyl trifluoromethanesulphonate ≥97.0% (by GC)
Catalog # TCIAC2326-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 97.0 %

Specification Test Results

About this item

 :
 :260.61 g/mol
Boiling Pt:120°C
 :GHS07
CAS Number:66107-36-6
Formula:C₇H₄ClF₃O₃S
MDL Number:MFCD07784322