Order Entry
Italy
ContactUsLinkComponent
cis-1,4-Cyclohexanediol ≥97.0% (by GC)
cis-1,4-Cyclohexanediol ≥97.0% (by GC)
Catalog # TCIAC2319-200MG
Supplier:  TCI
CAS Number:  
cis-1,4-Cyclohexanediol ≥97.0% (by GC)
Catalog # TCIAC2319-200MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    200 mg
  • Seq:
    2530

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 97.0 %
  • Melting point:
    108.0 to 114.0 deg-C

Specification Test Results

About this item

 :
 :116.16 g/mol
EINECS:213-240-8
CAS Number:931-71-5
Formula:C₆H₁₂O₂
MDL Number:MFCD00075462