


(R)-4-Chloromethyl-2,2-dimethyl-1,3-dioxolane ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:57044-24-3
:
Formula:C₆H₁₁ClO₂
MDL Number:MFCD00273371
:150.60 g/mol
Boiling Pt:63°C
Flash Pt:51°C
UN:1992
ADR:3,III
:GHS02
:GHS05
:GHS06
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
- Specific rotation [a]20/D:+40.0 to +48.0 deg (neat)
Specification Test Results
Frequently Bought Together