trans-1,4-Cyclohexanedimethanol ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:3236-48-4
:
Formula:C₈H₁₆O₂
MDL Number:MFCD00066360
:144.21 g/mol
Boiling Pt:164°C
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:64.0 to 68.0 deg-C
Specification Test Results
Frequently Bought Together