
2-(4-Chlorophenyl)-2-methylpropanol ≥98.0% (by GC)
Catalog # TCIAC2176-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:48.0 to 52.0 deg-C
Specification Test Results
About this item
:
:184.66 g/mol
:GHS07
CAS Number:80854-14-4
Formula:C₁₀H₁₃ClO
MDL Number:MFCD06657569