
Carbon nanotube multi-walled, Length: 1-2 µm, Ø 20-40 nm
Catalog # TCIAC2155-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Black powder to crystal
- Purity (TPO Method):min. 95 %
- Diameter:20 to 40 nm
- Length:1 to 2 micro_m
Specification Test Results
About this item
:
:12.011 g/mol
:GHS07
CAS Number:308068-56-6
Formula:C
MDL Number:MFCD00133992