
4-(4-Chlorophenyl)morpholine ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:70291-67-7
:
Formula:C₁₀H₁₂ClNO
MDL Number:MFCD09038510
ADR:6.1,III
:197.66 g/mol
UN:2811
:GHS06
Specification Test Results
- Purity (GC):min. 98.0 %
- Melting point:71.0 to 75.0 deg-C
Specification Test Results
Frequently Bought Together