Order Entry
Italy
ContactUsLinkComponent
18-Crown-5 [4-(2,4-dinitrophenylazo)phenol]
18-Crown-5 [4-(2,4-dinitrophenylazo)phenol]
Catalog # TCIAC1943-100MG
Supplier:  TCI
CAS Number:  
18-Crown-5 [4-(2,4-dinitrophenylazo)phenol]
Catalog # TCIAC1943-100MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Amber to Dark green powder to crystal

Specification Test Results

About this item

 :
 :506.47 g/mol
CAS Number:81238-58-6
Formula:C₂₂H₂₆N₄O₁₀