
cis-1,4-Cyclohexanediamine dihydrochloride ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:2121-79-1
:
Formula:C₆H₁₄N₂
MDL Number:MFCD20487918
:187.11 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together