Chloromethyl polystyrene resin 0.8 - 1.2 mmol/g, cross-linked with 2% DVB (100-200 mesh)
Supplier: TCI
Certificates
About this item
CAS Number:55844-94-5
:
MDL Number:MFCD00146406
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Loading:0.8 to 1.2 mmol/g
Specification Test Results
Frequently Bought Together