Order Entry
Italy
ContactUsLinkComponent
Chloromethyl polystyrene resin 0.8-1.2 mmol/g, cross-linked with 2% DVB (200-400 mesh)
Chloromethyl polystyrene resin 0.8-1.2 mmol/g, cross-linked with 2% DVB (200-400 mesh)
Codice catalogo TCIAC1746-5G
Supplier:  TCI
CAS Number:  
Chloromethyl polystyrene resin 0.8-1.2 mmol/g, cross-linked with 2% DVB (200-400 mesh)
Codice catalogo TCIAC1746-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Loading:
    0.8 to 1.2 mmol/g

Specification Test Results

About this item

 :
CAS Number:55844-94-5
MDL Number:MFCD00146406