
5-Chlorothiophene-2-carboxaldehyde ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:7283-96-7
:
Formula:C₅H₃ClOS
MDL Number:MFCD00047090
:146.59 g/mol
Flash Pt:97°C
EINECS:230-708-7
:GHS07
Specification Test Results
- Appearance:Colorless to Brown clear liquid
- Purity (GC):min. 95.0 %
Specification Test Results
Frequently Bought Together