Diethyl-trans-4-cyclohexene-1,2-dicarboxylate ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:5048-50-0
:
Formula:C₁₂H₁₈O₄
MDL Number:MFCD06799457
:226.27 g/mol
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together