
2-(Cyclohexylaminomethyl)pyridine ≥98.0% (by GC, titration analysis)
Codice catalogo TCIAC1685-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Light orange to Yellow to Green clear liquid
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
About this item
:
:190.29 g/mol
:GHS07
CAS Number:68339-45-7
Formula:C₁₂H₁₈N₂
MDL Number:MFCD01135534