
4-(Chloromethyl)thiazole hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:7709-58-2
:
Formula:C₄H₄ClNS
MDL Number:MFCD02094738
:170.05 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Argentmetric Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together