



(+)-B-Chlorodiisopinocampheylborane 58% ∼1.6 mol/L in hexane
Catalog # TCIAC1614-100ML
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Concentration (Argentmetric titration):58.0 to 64.0 w/w%
- Specific rotation [a]20/D:+47.0 to +54.0 deg (Calcd. on Hexane solution)
Specification Test Results
About this item
:
:320.75 g/mol
Density:0.88
Storage Temperature:Refrigerator
UN:2924
ADR:3,II
:GHS02
:GHS05
:GHS07
:GHS08
CAS Number:112246-73-8
Formula:C₂₀H₃₄BCl
MDL Number:MFCD00074808