
N-Carbobenzoxy-1,6-diaminohexane hydrochloride ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:78618-06-1
:
Formula:C₁₄H₂₂N₂O₂
MDL Number:MFCD00270151
:286.80 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:175.0 to 179.0 deg-C
Specification Test Results
Frequently Bought Together