



Cyclohexylmagnesium bromide ∼18% ∼1 mol/L in THF
Catalog # TCIAC1504-100G
Supplier: TCI
CAS Number:
Specifications
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- Concentration (sec Butanol method):18.0 to 19.0%
Specification Test Results
About this item
:
:187.36 g/mol
UN:2924
ADR:3,II
:GHS02
:GHS05
:GHS07
:GHS08
CAS Number:931-50-0
Formula:C₆H₁₁BrMg
MDL Number:MFCD01321152