



Tebbe Reagent ∼0.5 mol/L in toluene
TCIAC1411-25ML
: TCI
:
- Pk:25 mL
- Seq:2315
- Appearance:Dark red clear liquid to cloudy liquid
- Concentration (Argentmetric titration):22.0 to 27.0 w/w%
:
:284.62 g/mol
:14,09091
:1°C
:Refrigerator
:2924
:3,II
:GHS02
:GHS05
:GHS07
:GHS08
:67719-69-1
:C₁₃H₁₈AlClTi
:MFCD00151575