Order Entry
Italy
ContactUsLinkComponent
4-Cholesten-3-one ≥95.0% (by GC)
4-Cholesten-3-one ≥95.0% (by GC)
Catalog # TCIAC1366-5G
Supplier:  TCI
CAS Number:  
4-Cholesten-3-one ≥95.0% (by GC)
Catalog # TCIAC1366-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (GC):
    min. 95.0 %
  • Melting point:
    79.0 to 83.0 deg-C
  • Specific rotation [a]20/D:
    +91.0 to +95.0 deg (C=2, CHCl3)

Specification Test Results

About this item

 :
 :384.65 g/mol
EINECS:210-005-1
CAS Number:601-57-0
Formula:C₂₇H₄₄O
MDL Number:MFCD00003663