Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Diethyl cyclopentylmalonate
Diethyl cyclopentylmalonate ≥95.0% (by GC)
Diethyl cyclopentylmalonate ≥95.0% (by GC)
TCIAC0944-25ML
:
TCI
:
Diethyl cyclopentylmalonate ≥95.0% (by GC)
TCIAC0944-25ML
:
TCI
:
Pk:
25 mL
Seq:
2315
Appearance:
Colorless to Light yellow clear liquid
Purity (GC):
min. 95.0 %
:
:
228.29 g/mol
:
138°C
:
18928-91-1
:
C₁₂H₂₀O₄
:
MFCD00059514