Order Entry
Italy
ContactUsLinkComponent
4,4-Dimethoxybutanenitrile ≥98.0% (by GC)
Irritant
4,4-Dimethoxybutanenitrile ≥98.0% (by GC)
Catalog # TCIAC0891-25ML
Supplier:  TCI
CAS Number:  
Irritant
4,4-Dimethoxybutanenitrile ≥98.0% (by GC)
Catalog # TCIAC0891-25ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :129.1570 g/mol
Boiling Pt:110°C
Flash Pt:91°C
EINECS:238-658-8
 :GHS07
CAS Number:14618-78-1
Formula:C₆H₁₁NO₂
MDL Number:MFCD00019883