Order Entry
Italy
ContactUsLinkComponent
3-Cyclohexene-1-carboxaldehyde ≥98.0% (by GC)
Flammable
Irritant
3-Cyclohexene-1-carboxaldehyde ≥98.0% (by GC)
Catalog # TCIAC0881-25ML
Supplier:  TCI
CAS Number:  
Flammable
Irritant
3-Cyclohexene-1-carboxaldehyde ≥98.0% (by GC)
Catalog # TCIAC0881-25ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
Boiling Pt:164°C
Flash Pt:50°C
Storage Temperature:Refrigerator
UN:2498
ADR:3,III
 :110.16 g/mol
EINECS:202-858-3
 :GHS02
 :GHS07
CAS Number:100-50-5
Formula:C₇H₁₀O
MDL Number:MFCD00001572