S-(Carboxymethyl)-L-cysteine ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAC0863-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Solubility in hot Water:almost transparency
Specification Test Results
About this item
:
:179.19 g/mol
Merck Index:14,01802
EINECS:211-327-5
CAS Number:638-23-3
Formula:C₅H₉NO₄S
MDL Number:MFCD00002614