Order Entry
Italy
ContactUsLinkComponent
 
 
1,1-Cyclopentanediacetic anhydride ≥98.0% (by GC, titration analysis)
  TCIAC0839-5G
 :  TCI
 :  
 
1,1-Cyclopentanediacetic anhydride ≥98.0% (by GC, titration analysis)
  TCIAC0839-5G
 :  TCI
 :  

 

  • Pk:
    5 g
  • Seq:
    3173

 

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Morpholine Method):
    min. 98.0 %
  • Melting point:
    65.0 to 68.0 deg-C

 

 

 :
 :168.19 g/mol
 :176°C
 :227-116-6
 :GHS07
 :5662-95-3
 :C₉H₁₂O₃
 :MFCD00006678