
5-Chloro-2-methylaniline Hydrochloride ≥98.0% (by HPLC, total nitrogen)
Catalog # TCIAC0746-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Light yellow to Light red powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Solubility in Methanol:almost transparency
Specification Test Results
About this item
:
:178.06 g/mol
EINECS:228-403-9
:GHS07
CAS Number:6259-42-3
Formula:C₇H₈ClN
MDL Number:MFCD00035487