Nε-carbobenzoxy-Nɑ-tosyl-L-lysine ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
Synonyms: Nepsilon-Cbz-Nalpha-tosyl-L-lysine, Tos-Lys(Z)-OH
CAS Number:34235-82-0
:
Formula:C₂₁H₂₆N₂O₆S
MDL Number:MFCD00059201
:434.51 g/mol
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 95.0 %
- Melting point:121.0 to 124.0 deg-C
- Specific rotation [a]20/D:+13.0 to +16.0 deg (C=1, EtOH)
Specification Test Results
Frequently Bought Together