
(+)-Catechin hydrate ≥97.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:225937-10-0
:154-23-4
Formula:C₁₅H₁₄O₆·xH₂O
MDL Number:MFCD00075649
:290.27 g/mol (anhy)
Merck Index:14,01902
Melting Pt:96°C
EINECS:205-825-1
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Yellow to Orange powder to crystal
- Purity (HPLC):min. 97.0 area%
- Specific rotation [a]20/D:+13.0 to +18.0 deg (C=5, Acetone (50%)) (calcd.on anh.substance)
- Water:3.0 to 5.0 %
Specification Test Results
Frequently Bought Together