Cholesterol Phenylacetate ≥94.0% (by GC)
Catalog # TCIAC0693-10G
Supplier: TCI
CAS Number:
Specifications
- Pk:10 g
- Seq:3236
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 94.0 %
- Mesomorphic range:119.0 to 126.0 deg-C
- Specific rotation [a]20/D:-20.0 to -26.0 deg (C=5, THF)
Specification Test Results
About this item
:
:504.80 g/mol
EINECS:251-781-1
CAS Number:33998-26-4
Formula:C₃₅H₅₂O₂
MDL Number:MFCD00021140