Cortisone acetate ≥99.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:50-04-4
:
Formula:C₂₃H₃₀O₆
MDL Number:MFCD00003609
:402.49 g/mol
Merck Index:14,02539
EINECS:200-006-5
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 99.0 area%
- Purity (UV-vis method):min. 98.0 %
- Specific rotation [a]20/D:+207 to +216 deg (C=1, MeOH)
- Solubility in hot Methanol:almost transparency
Specification Test Results
Frequently Bought Together