Order Entry
Italy
ContactUsLinkComponent
 
ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na, oxidized form) ≥90.0% (by HPLC) for biochemical research
 :  TCI
 
ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na, oxidized form) ≥90.0% (by HPLC) for biochemical research
 :  TCI

 

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (HPLC):
    min. 90.0 area%
  • abs-ratio (a/b):
    0.80 to 0.86 (E250/E260)
  • abs-ratio (2a/2b):
    0.19 to 0.23 (E280/E260)
  • Absorbance (E1%1cm):
    min. 200 (258.0 to 262.0 nm, pH7.5)

 

Frequently Bought Together