


1-Chloro-2-propanol ≥70% (by GC) (contains ca. 25% 2-Chloro-1-propanol)
Supplier: TCI
Certificates
About this item
CAS Number:127-00-4
:
Formula:C₃H₇ClO
MDL Number:MFCD00004530
:94.54 g/mol
Merck Index:14,07851
Boiling Pt:132°C
Flash Pt:52°C
EINECS:204-819-6
UN:2611
ADR:6.1,II
:GHS02
:GHS06
:GHS08
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 70.0 %
Specification Test Results
Frequently Bought Together