Order Entry
Italy
ContactUsLinkComponent
 
Chlorogenic acid hydrate ≥98.0% (by HPLC, titration analysis)
 :  TCI
 
undefined
Chlorogenic acid hydrate ≥98.0% (by HPLC, titration analysis)
 :  TCI

 

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Neutralization titration):
    min. 98.0 % (calcd.on anh.substance)
  • Specific rotation [a]20/D:
    -35.0 to -38.0 deg (C=1, H2O)
  • Solubility in hot Water:
    almost transparency

 

Frequently Bought Together