3'-Chloro-4-dimethylaminoazobenzene ≥97.0% (by titrimetric analysis)
TCIAC0158-25G
: TCI
:
- Pk:25 g
- Seq:3308
- Appearance:Orange to Amber to Dark red powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):gt. 97.0 %
- Melting point:101.0 to 105.0 deg-C
- Solubility in hot Methanol:very faint turbidity
:
:259.74 g/mol
:3789-77-3
:C₁₄H₁₄ClN₃
:MFCD00059480