
3-Benzo[b]thiophenesulphonyl chloride ≥95.0% (by GC, titration analysis)
Catalog # TCIAB6229-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystaline
- Purity (GC):min. 95.0 %
- Purity (Argentmetric Titration):min. 95.0 %
- Melting point:85.0 to 89.0 deg-C
Specification Test Results
About this item
:
:232.71 g/mol
Boiling Pt:142°C
UN:3261
ADR:8,II
:GHS05
CAS Number:18494-87-6
Formula:C₈H₅ClO₂S₂
MDL Number:MFCD02681897