Order Entry
Italy
ContactUsLinkComponent
 
2-Bromo-3-(2-octyldodecyl)thiophene ≥95.0% (by GC)
  TCIAB6166-1G
 :  TCI
 :  
2-Bromo-3-(2-octyldodecyl)thiophene ≥95.0% (by GC)
  TCIAB6166-1G
 :  TCI
 :  

 

  • Pk:
    1 g
  • Seq:
    3047

 

 

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 95.0 %

 

 

 :
 :443.57 g/mol
 :1268060-77-0
 :C₂₄H₄₃BrS
 :MFCD27901946