4,4'-Bipyridine -1,1'-dioxide ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:24573-15-7
:
Formula:C₁₀H₈N₂O₂
MDL Number:MFCD00134383
:188.19 g/mol
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together