Order Entry
Italy
ContactUsLinkComponent
Dimethyl-7,7'-dimethoxy[4,4'-bibenzo[d][1,3]dioxole]5,5'-dicarboxylate ≥98.0% (by GC)
Dimethyl-7,7'-dimethoxy[4,4'-bibenzo[d][1,3]dioxole]5,5'-dicarboxylate ≥98.0% (by GC)
Supplier:  TCI
Certificates
Dimethyl-7,7'-dimethoxy[4,4'-bibenzo[d][1,3]dioxole]5,5'-dicarboxylate ≥98.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    179.0 to 183.0 deg-C

Specification Test Results

Frequently Bought Together