1,4-Bis(4-phenoxyphenoxy)benzene ≥97.0% (by HPLC)
Catalog # TCIAB6098-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 97.0 area%
Specification Test Results
About this item
:
Boiling Pt:321°C
Melting Pt:151°C
:446.50 g/mol
CAS Number:2455-43-8
Formula:C₃₀H₂₂O₄